Assay Details
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012266.1 | - | 2 - 3 | 463 | 110 | |
AK027977.1 | - | 2 - 3 | 435 | 110 | ||
BC004620.1 | - | 2 - 3 | 441 | 3585323 | 110 | |
BC004703.1 | - | 2 - 3 | 1073 | 3499265 | 110 | |
X97755.1 | - | 2 - 3 | 439 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
