Assay Details
Gene Symbol: Sec22c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178677.4 | NP_848792.2 | 1 - 2 | 105 | 111 | |
GenBank mRNA | BB661643.1 | - | 1 - 2 | 105 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_178677.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
