Assay Details
Gene Symbol: Sec22c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178677.4 | NP_848792.2 | 2 - 3 | 228 | 68 | |
GenBank mRNA | AK029985.1 | - | 2 - 3 | 159 | 68 | |
BB661643.1 | - | 2 - 3 | 228 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK029985.1, NM_178677.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
