Assay Details
| Gene Symbol: Gm12530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040669.1 | 2 - 3 | 228 | 108 | ||
| GenBank mRNA | AK052790.1 | - | 2 - 3 | 229 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | erythrocyte protein band 4.1 like 4b | Epb4.1l4b |
6430543G08Rik, AA589614, AU021054, BB007528, D4Ertd346e, Ehm2, Epb41l4b, Lulu2 |
-30.0kb | Anti-sense | Chr. 4 - 56992043 - 57143168 on NCBI Build GRCm38 |
| Downstream |
protein tyrosine phosphatase, non-receptor type 3 |
Ptpn3 |
9530011I20Rik, PTP-H1, PTPCL |
16.1kb | Anti-sense | Chr. 4 - 57190841 - 57339046 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12530
Genomic Map