Assay Details
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021332.2 | NP_067307.2 | 7 - 8 | 837 | 94 | |
GenBank mRNA | AJ001692.1 | - | 7 - 8 | 827 | 94 | |
BC103624.1 | - | 7 - 8 | 826 | 94 | ||
BC105649.1 | - | 7 - 8 | 826 | 94 | ||
BC105650.1 | - | 7 - 8 | 826 | 94 | ||
BC139464.1 | - | 7 - 8 | 864 | 8862486 | 94 | |
BC139466.1 | - | 7 - 8 | 864 | 8862488 | 94 | |
BC145626.1 | - | 7 - 8 | 864 | 9054194 | 94 | |
DQ093397.1 | - | 7 - 8 | 837 | 94 | ||
DQ093398.1 | - | 7 - 8 | 837 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
