Assay Details
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024922.2 | NP_001020093.2 | 10 - 11 | 1118 | 89 | |
GenBank mRNA | AK191777.1 | - | 2 - 3 | 92 | 89 | |
BC085301.1 | - | 10 - 11 | 1192 | 89 | ||
BC099554.1 | - | 10 - 11 | 1125 | 30308124 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950972 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950969 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950970 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950968 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950971 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541975 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386867 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712928 | AK046101.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712928 | AK083681.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712928 | BC056462.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712928 | NM_001029868.1 | refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109612 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | BC085301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | BC099554.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | BC085301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | BC099554.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950957 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950972 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01762854 | - | - |
Whole Mouse Genome G4122A | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P159885 | BC085301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P159885 | BC099554.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P159885 | NM_001024922.2 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07066665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07066664 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07066668 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950972 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950969 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950968 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950971 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25950970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06907986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07316893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07316888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06753274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07075651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06929775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25950961 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10579233 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579232 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579225 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579228 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579230 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579229 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579231 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579227 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579223 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579221 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579224 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579226 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579222 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579220 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435681_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455563_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457947_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10572345 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10579219 | - | - |
Mouse Genome 430 2.0 Array | 1435681_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1435681_s_at | - | - |
Mouse Genome 430 2.0 Array | 1455563_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457947_at | - | - |
Mouse Genome 430 2.0 Array | 1457947_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455563_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234374_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234374_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800018044.mm.1 | NM_001024922.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018044.mm.1 | NM_001024922.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018044.mm.1 | TC0800002389.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800018042.mm.1 | NM_001024922.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018042.mm.1 | NM_001024922.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018042.mm.1 | TC0800002389.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800018040.mm.1 | NM_001024922.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018040.mm.1 | NM_001024922.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018040.mm.1 | TC0800002389.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800018041.mm.1 | NM_001024922.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018041.mm.1 | NM_001024922.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018041.mm.1 | TC0800002389.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800018043.mm.1 | NM_001024922.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018043.mm.1 | NM_001024922.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800018043.mm.1 | TC0800002389.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435681_PM_s_at | - | - |
Mouse Expression Set 430 | 1455563_PM_at | - | - |
Mouse Expression Set 430 | 1455563_at | - | - |
Mouse Expression Set 430 | 1435681_s_at | - | - |
Mouse Expression Set 430 | 1457947_at | - | - |
Mouse Expression Set 430 | 1457947_PM_at | - | - |
Murine Genome U74v2 Set | 102015_PM_at | - | - |
Murine Genome U74v2 Set | 102015_at | - | - |
Mouse Genome 430A 2.0 Array | 1435681_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1455563_at | - | - |
Mouse Genome 430A 2.0 Array | 1455563_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435681_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455563_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455563_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435681_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435681_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457947_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457947_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 584563 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237785 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2801380 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2801380 | - | - |
