Assay Details
Gene Symbol: Gm20756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081540.1 | - | 1 - 2 | 1429 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protection of telomeres 1A | Pot1a |
1500031H18Rik, AI851169, Pot1 |
-145.2kb | Anti-sense | Chr. 6 - 25743735 - 25809277 on NCBI Build GRCm38 |
Downstream |
glutamate receptor, metabotropic 8 |
Grm8 |
A230002O04, Gprc1h, mGluR8 |
1.3mb | Anti-sense | Chr. 6 - 27275121 - 28135837 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
