Assay Details
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033397.5 | NP_001028569.2 | 7 - 8 | 1298 | 62 | |
GenBank mRNA | AK028591.1 | - | 7 - 8 | 1298 | 62 | |
AK028682.1 | - | 6 - 7 | 2431 | 62 | ||
AK132445.1 | - | 7 - 8 | 1302 | 62 | ||
AK162718.1 | - | 7 - 8 | 1300 | 62 | ||
BC116672.1 | - | 7 - 8 | 1287 | 40107596 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK162718.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK132445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK028591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK162718.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK132445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | AK028591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556811 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364590 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | AB041611.2 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | AK083905.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | AK128943.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | AK147847.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | BC014713.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | L19622.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | L27424.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | NM_011595.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704638 | Z30970.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66179 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100822 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK162718.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK132445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK028591.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK162718.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK132445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | AK028591.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07460898 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02408005 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07460887 | - | - |
Whole Mouse Genome G4122A | A_51_P501873 | AK162718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | AK132445.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | AK028591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | AK162718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | AK132445.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | AK028591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501873 | NM_001033397.5 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07460887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09116578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09406534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09406532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09433311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09451565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09381061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09608842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09433312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09516518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061717 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390814 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390813 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390807 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390810 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390808 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390809 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390812 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390815 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390811 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436160_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390806 | - | - |
Mouse Genome 430 2.0 Array | 1436160_at | - | - |
Mouse Genome 430 2.0 Array | 1436160_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320864_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320864_PM_at | - | - |
Mouse Expression Set 430 | 1436160_PM_at | - | - |
Mouse Expression Set 430 | 1436160_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436160_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436160_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2765981 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2765981 | - | - |
