Assay Details
| Gene Symbol: A930013N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020852.1 | - | 1 | 847 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP-ribosylation factor-like 6 | Arl6 |
1110018H24Rik, 2210411E14Rik, BBS3 |
Sense | Chr. 16 - 59613321 - 59639405 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930547E14 gene | 4930547E14Rik | 20.0kb | Anti-sense | Chr. 16 - 59636945 - 59672993 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930013N22Rik
Genomic Map