Assay Details
Gene Symbol: Cfap57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021161.1 | - | 1 | 1147 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 125 | Tmem125 |
6330530A05Rik, AV223516 |
-24.1kb | Anti-sense | Chr. 4 - 118540941 - 118544953 on NCBI Build GRCm38 |
Downstream | EBNA1 binding protein 2 | Ebna1bp2 |
1810014B19Rik, AA537058, B830003A16Rik, C81321, Ebp2, Nobp, p40 |
50.4kb | Sense | Chr. 4 - 118620799 - 118627776 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
