Assay Details
Gene Symbol: F730016J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090172.1 | - | 1 - 2 | 120 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ficolin B | Fcnb | Fcn2 | -33.3kb | Anti-sense | Chr. 2 - 28076479 - 28084913 on NCBI Build GRCm38 |
Downstream | olfactomedin 1 | Olfm1 |
AMY, AW742568, Noe1, OlfA, Pancortin, Pancortin3 |
71.4kb | Sense | Chr. 2 - 28193093 - 28230736 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
