Assay Details
Gene Symbol: Gm13446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045894.1 | 1 - 2 | 250 | 91 | ||
GenBank mRNA | AK138201.1 | - | 1 - 2 | 251 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
