Assay Details
| Gene Symbol: Gm13269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146025.1 | - | 1 - 2 | 242 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10848 | Gm10848 | ENSMUSG00000075523 | -6.2kb | Sense | Chr. 2 - 14709329 - 14739758 on NCBI Build GRCm38 |
| Downstream | NOL1/NOP2/Sun domain family member 6 | Nsun6 |
4933403D21Rik, 4933414E04Rik, NOPD1 |
228.0kb | Sense | Chr. 2 - 14991522 - 15054872 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13269
Genomic Map