Assay Details
Gene Symbol: Tmem91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290497.1 | NP_001277426.1 | 2 - 3 | 373 | 100 | |
NM_001290498.1 | NP_001277427.1 | 2 - 3 | 373 | 100 | ||
NM_177102.4 | NP_796076.1 | 2 - 3 | 373 | 100 | ||
GenBank mRNA | AK043858.1 | - | 2 - 3 | 326 | 100 | |
AK079312.1 | - | 2 - 3 | 374 | 100 | ||
BC058996.1 | - | 2 - 3 | 322 | 5711063 | 100 | |
BE863748.1 | - | 1 - 2 | 254 | 100 | ||
BQ555196.1 | - | 2 - 3 | 301 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
