Assay Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019870.3 | NP_063923.1 | 7 - 8 | 639 | 82 | |
| GenBank mRNA | AJ627039.1 | - | 6 - 7 | 458 | 82 | |
| AK014469.1 | - | 7 - 8 | 568 | 82 | ||
| AY183136.1 | - | 7 - 8 | 608 | 82 | ||
More Information
Set Membership: |
|

Naa10
Genomic Map