Assay Details
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009025.2 | NP_033051.2 | 1 - 2 | 199 | 63 | |
GenBank mRNA | AK053353.1 | - | 1 - 2 | 196 | 63 | |
AK141511.1 | - | 1 - 2 | 206 | 63 | ||
AK147896.1 | - | 1 - 2 | 167 | 63 | ||
AK166354.1 | - | 1 - 2 | 196 | 63 | ||
BC057300.1 | - | 1 - 2 | 190 | 6821254 | 63 | |
BC060195.1 | - | 1 - 2 | 215 | 63 | ||
BC068297.1 | - | 1 - 2 | 63 | 30102545 | 63 | |
CA749631.1 | - | 1 - 2 | 211 | 63 | ||
U20238.1 | - | 1 - 2 | 184 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
