Assay Details
Gene Symbol: 1810034E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045798.1 | 1 - 2 | 503 | 93 | ||
GenBank mRNA | AK002708.1 | - | 1 - 2 | 151 | 93 | |
AK143964.1 | - | 1 - 2 | 458 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hyaluronic acid binding protein 4 | Habp4 |
4933413D03Rik, 4933428J01Rik |
-62.2kb | Sense | Chr. 13 - 64161866 - 64186537 on NCBI Build GRCm38 |
Downstream | AhpC/TSA antioxidant enzyme domain containing 1 | Aaed1 |
1110018J18Rik, AI849003 |
25.3kb | Anti-sense | Chr. 13 - 64291836 - 64312710 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700000027.rn
Additional Information:
This assay is in the 5'UTR region of the AK143964.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
