Assay Details
Gene Symbol: PGRMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308580.1 | NP_001295509.1 | 2 - 3 | 560 | 106 | |
GenBank mRNA | KM114208.1 | - | 2 - 3 | 560 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
