Assay Details
| Gene Symbol: ST6GALNAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005616.4 | XP_004005665.1 | 6 - 7 | 935 | 79 | |
| XM_012162328.3 | XP_012017718.1 | 5 - 6 | 882 | 79 | ||
| XM_012162341.3 | XP_012017731.1 | 5 - 6 | 1054 | 79 | ||
| XM_027966408.1 | XP_027822209.1 | 6 - 7 | 1297 | 79 | ||
| XM_027966409.1 | XP_027822210.1 | 6 - 7 | 979 | 79 | ||
| XM_027966410.1 | XP_027822211.1 | 6 - 7 | 1099 | 79 | ||
| XM_027966411.1 | XP_027822212.1 | 5 - 6 | 903 | 79 | ||
| XM_027966412.1 | XP_027822213.1 | 5 - 6 | 858 | 79 | ||
| XM_027966413.1 | XP_027822214.1 | 4 - 5 | 603 | 79 | ||
| XM_027966414.1 | XP_027822215.1 | 5 - 6 | 573 | 79 | ||
More Information
Set Membership: |
|

ST6GALNAC6
Genomic Map