Assay Details
Gene Symbol: NSUN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009785.4 | XP_004009834.3 | 11 - 12 | 1722 | 76 | |
XM_027970950.1 | XP_027826751.1 | 11 - 12 | 1722 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
