Assay Details
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016872.3 | XP_004016921.1 | 5 - 6 | 1731 | 82 | |
XM_012096729.3 | XP_011952119.1 | 5 - 6 | 1893 | 82 | ||
XM_012096730.3 | XP_011952120.1 | 5 - 6 | 2329 | 82 | ||
XM_027979995.1 | XP_027835796.1 | 3 - 4 | 1957 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
