Assay Details
Gene Symbol: TEX261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094446.2 | XP_014949932.1 | 2 - 3 | 252 | 75 | |
XM_027966973.1 | XP_027822774.1 | 2 - 3 | 227 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
