Assay Details
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171288.1 | NP_001164759.1 | 4 - 5 | 569 | 80 | |
XM_008261390.2 | XP_008259612.1 | 6 - 7 | 799 | 80 | ||
XM_008261391.2 | XP_008259613.1 | 6 - 7 | 774 | 80 | ||
XM_008261392.2 | XP_008259614.1 | 6 - 7 | 770 | 80 | ||
XM_008261393.2 | XP_008259615.1 | 6 - 7 | 746 | 80 | ||
XM_008261394.2 | XP_008259616.1 | 5 - 6 | 1254 | 80 | ||
XM_017343656.1 | XP_017199145.1 | 6 - 7 | 1242 | 80 | ||
XM_017343657.1 | XP_017199146.1 | 6 - 7 | 1217 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
