Assay Details
Gene Symbol: LOC4327063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766457.2 | XP_015621943.1 | 1 - 2 | 632 | 144 | |
GenBank mRNA | AK068290.1 | - | 1 - 2 | 616 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
