Assay Details
Gene Symbol: LOC4328877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771644.2 | XP_015627130.1 | 3 - 4 | 1077 | 101 | |
GenBank mRNA | AK243018.1 | - | 3 - 4 | 1061 | 101 | |
AY429650.1 | - | 3 - 4 | 1066 | 101 | ||
EF122464.1 | - | 3 - 4 | 1050 | 101 | ||
JQ694716.1 | - | 3 - 4 | 999 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
