Assay Details
| Gene Symbol: LOC4331296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776597.2 | XP_015632083.1 | 37 - 38 | 5515 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4331296| Gene Symbol: LOC4331296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776597.2 | XP_015632083.1 | 37 - 38 | 5515 | 79 | |
Set Membership: |
|