Assay Details
Gene Symbol: LOC4334782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772693.2 | XP_015628179.1 | 4 - 5 | 929 | 82 | |
XM_015772694.2 | XP_015628180.1 | 4 - 5 | 897 | 82 | ||
GenBank mRNA | AK065662.1 | - | 3 - 4 | 886 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
