Assay Details
| Gene Symbol: LOC4339711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784070.1 | XP_015639556.1 | 1 | 945 | 85 | |
| GenBank mRNA | AK243635.1 | - | 1 | 951 | 85 | |
More Information
Set Membership: |
|

LOC4339711
Genomic Map