Assay Details
Gene Symbol: LOC4343805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789589.2 | XP_015645075.1 | 1 - 2 | 387 | 113 | |
GenBank mRNA | AK073214.1 | - | 1 - 2 | 364 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
