Assay Details
Gene Symbol: LOC4338246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782516.2 | XP_015638002.1 | 1 - 2 | 159 | 63 | |
GenBank mRNA | AK065555.1 | - | 1 - 2 | 160 | 63 | |
AK065811.1 | - | 1 - 2 | 160 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782516.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
