Assay Details
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257814.2 | NP_001244743.1 | 469 | 72 | ||
XM_015126605.2 | XP_014982091.1 | 490 | 72 | |||
GenBank mRNA | JU335580.1 | - | 469 | 72 | ||
JU476491.1 | - | 469 | 72 | |||
JV047726.1 | - | 474 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
