Assay Details
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261059.2 | NP_001247988.1 | 383 | 150 | ||
XM_015134892.2 | XP_014990378.1 | 1056 | 150 | |||
GenBank mRNA | JU319855.1 | - | 287 | 150 | ||
JV048144.1 | - | 384 | 150 | |||
JV660459.1 | - | 660 | 150 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
