Assay Details
Gene Symbol: MICB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114956.1 | NP_001108428.1 | 1032 | 86 | ||
GenBank mRNA | AF055388.1 | - | 1014 | 86 | ||
AF322860.1 | - | 1020 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
