Assay Details
| Gene Symbol: EIF2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266005.1 | NP_001252934.1 | 484 | 64 | ||
| XM_015144118.1 | XP_014999604.1 | 550 | 64 | |||
| XM_015144119.2 | XP_014999605.1 | 520 | 64 | |||
| GenBank mRNA | JU321791.1 | - | 519 | 64 | ||
| JU477678.1 | - | 484 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EIF2B2
Genomic Map