Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260683.1 | NP_001247612.1 | 512 | 135 | ||
XM_028844810.1 | XP_028700643.1 | 785 | 135 | |||
GenBank mRNA | JU333209.1 | - | 493 | 135 | ||
JU474525.1 | - | 512 | 135 | |||
JV644046.1 | - | 529 | 135 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
