Assay Details
| Gene Symbol: VKORC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266931.2 | NP_001253860.1 | 305 | 108 | ||
| GenBank mRNA | JU474461.1 | - | 309 | 108 | ||
| JV046286.1 | - | 305 | 108 | |||
| JV635978.1 | - | 307 | 108 | |||
| JV674346.1 | - | 315 | 108 | |||
More Information
Set Membership: |
|

VKORC1L1
Genomic Map