Assay Details
Gene Symbol: LGALS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168627.1 | NP_001162098.1 | 155 | 114 | ||
GenBank mRNA | EU152916.1 | - | 155 | 114 | ||
JV044013.1 | - | 180 | 114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
