Assay Details
| Gene Symbol: LOC705696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125931.2 | XP_014981417.1 | 2523 | 69 | ||
| XM_015125932.2 | XP_014981418.1 | 2493 | 69 | |||
| XM_028841348.1 | XP_028697181.1 | 2444 | 69 | |||
| GenBank mRNA | JU320447.1 | - | 2450 | 69 | ||
| JV044047.1 | - | 2369 | 69 | |||
| JV524154.1 | - | 2463 | 69 | |||
| JV725930.1 | - | 559 | 69 | |||
More Information
Set Membership: |
|

LOC705696
Genomic Map