Assay Details
Gene Symbol: Sftpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017342.2 | NP_059038.1 | 1 - 2 | 57 | 78 | |
GenBank mRNA | BC072693.1 | - | 1 - 2 | 50 | 7099876 | 78 |
X14221.1 | - | 1 - 2 | 58 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
