Assay Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053582.2 | NP_446034.1 | 9 - 10 | 1218 | 60 | |
| XM_006238977.2 | XP_006239039.1 | 10 - 11 | 1310 | 60 | ||
| XM_006238978.2 | XP_006239040.1 | 10 - 11 | 1307 | 60 | ||
| GenBank mRNA | AB050717.1 | - | 9 - 10 | 1219 | 60 | |
| BC097299.1 | - | 10 - 11 | 1277 | 7445067 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tinagl1
Genomic Map