Assay Details
Gene Symbol: Atp5i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080481.1 | NP_536729.1 | 2 - 3 | 104 | 82 | |
GenBank mRNA | BC058449.1 | - | 102 | 6890563 | 82 | |
D13121.1 | - | 2 - 3 | 104 | 82 | ||
FQ223740.1 | - | 2 - 3 | 94 | 82 | ||
FQ224619.1 | - | 243 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
