Assay Details
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002819.2 | NP_001002819.2 | 17 - 18 | 1945 | 79 | |
GenBank mRNA | BC098630.1 | - | 17 - 18 | 1945 | 7105780 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97186 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18793 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18793 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18793 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18793 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99267 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13695 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18686 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20843 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10733154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733159 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10733139 | - | - |
Rat Genome 230 2.0 Array | 1377761_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377761_at | - | - |
Affymetrix HT Rat Focus Array | 1377761_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377761_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000004153.rn.1 | TC1000000507.rn.1 | RTA TC |
Rat Expression Set 230 | 1377761_at | - | - |
Rat Expression Set 230 | 1377761_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733139 | - | - |
Rat Genome U34 Set | rc_AI045026_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21462573 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360656 | - | - |
