Assay Details
Gene Symbol: DKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039749.1 | NP_001034838.1 | 6 - 7 | 862 | 89 | |
XM_005661123.3 | XP_005661180.2 | 1033 | 89 | |||
XM_021082083.1 | XP_020937742.1 | 951 | 89 | |||
GenBank mRNA | AB232525.1 | - | 6 - 7 | 862 | 89 | |
AK392236.1 | - | 7 - 8 | 991 | 89 | ||
AK400778.1 | - | 7 - 8 | 987 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
