Assay Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244956.1 | NP_001231885.1 | 9 - 10 | 1408 | 67 | |
XM_021091035.1 | XP_020946694.1 | 1445 | 67 | |||
XM_021091036.1 | XP_020946695.1 | 1439 | 67 | |||
XM_021091037.1 | XP_020946696.1 | 2668 | 67 | |||
GenBank mRNA | AK234175.1 | - | 6 - 7 | 696 | 67 | |
AK234396.1 | - | 9 - 10 | 1404 | 67 | ||
AK400305.1 | - | 9 - 10 | 1423 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
