Assay Details
Gene Symbol: MRPL44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243405.1 | NP_001230334.1 | 3 - 4 | 844 | 90 | |
GenBank mRNA | CK467514.1 | - | 2 - 3 | 223 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
