Assay Details
Gene Symbol: LOC100525745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126168.5 | XP_003126216.4 | 969 | 91 | ||
GenBank mRNA | AK349605.1 | - | 4 - 5 | 952 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
