Assay Details
Gene Symbol: hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008673.1 | NP_001008673.1 | 7 - 8 | 738 | 66 | |
XM_018092512.1 | XP_017948001.1 | 7 - 8 | 777 | 66 | ||
GenBank mRNA | BC080462.1 | - | 7 - 8 | 738 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
