Genomic Map
LOADING...
Displaying...

Build 1.0 Frog Chr.Un|NW_003163916.1: Xt03697413_g1

Assay Details
Gene Symbol: ptgr1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011193.2 | NP_001011193.1 | 3 - 4 | 278 | 94 | |
XM_018092841.1 | XP_017948330.1 | 3 - 4 | 336 | 94 | ||
GenBank mRNA | BC087566.2 | - | 3 - 4 | 278 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
