Assay Details
Gene Symbol: mrps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113930.1 | NP_001107402.1 | 2 - 3 | 252 | 73 | |
XM_012971520.2 | XP_012826974.1 | 2 - 3 | 282 | 73 | ||
GenBank mRNA | BC157364.1 | - | 2 - 3 | 252 | 73 | |
CU075466.1 | - | 2 - 3 | 259 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
