Assay Details
Gene Symbol: LOC100285793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158683.1 | NP_001152155.1 | 402 | 57 | ||
GenBank mRNA | EU973974.1 | - | 402 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
